Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp4r1l-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027957.1 | 11 - 12 | 2131 | 90 | ||
GenBank mRNA | AK029171.1 | - | 12 - 13 | 4411 | 90 | |
AK035093.1 | - | 3 - 4 | 1772 | 90 | ||
AK041597.1 | - | 4 - 5 | 1369 | 90 | ||
AK046224.1 | - | 10 - 11 | 2028 | 90 | ||
AK047187.1 | - | 3 - 4 | 1222 | 90 | ||
AK161459.1 | - | 10 - 11 | 2234 | 90 | ||
AK164611.1 | - | 11 - 12 | 2133 | 90 | ||
AK170078.1 | - | 10 - 11 | 1215 | 90 | ||
BC064312.1 | - | 3 - 4 | 597 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |