Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176940.5 | NP_795914.3 | 1 - 2 | 299 | 63 | |
GenBank mRNA | AK038792.1 | - | 1 - 2 | 289 | 63 | |
AK087622.1 | - | 1 - 2 | 300 | 63 | ||
AK162888.1 | - | 1 - 2 | 293 | 63 | ||
BC082552.1 | - | 1 - 2 | 239 | 63 | ||
BY731616.1 | - | 1 - 2 | 296 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |