Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176940.5 | NP_795914.3 | 3 - 4 | 505 | 95 | |
GenBank mRNA | AK087622.1 | - | 3 - 4 | 506 | 95 | |
AK162888.1 | - | 3 - 4 | 499 | 95 | ||
BC082552.1 | - | 3 - 4 | 445 | 95 | ||
BC144786.1 | - | 1 - 2 | 95 | 9053334 | 95 | |
BC144787.1 | - | 1 - 2 | 95 | 95 | ||
BC145703.1 | - | 1 - 2 | 95 | 40131006 | 95 | |
BY731616.1 | - | 3 - 4 | 502 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |