Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm15894 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK038870.1 | - | 405 | 92 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | metastasis suppressor 1-like | Mtss1l | ABBA | -14.2kb | Sense | Chr. 8 - 110721282 - 110741400 on NCBI Build GRCm38 |
Downstream |
splicing factor 3b, subunit 3 |
Sf3b3 |
1810061H24Rik, 5730409A01Rik, AA409318, D8Ertd633e, RSE1, SAP130, mKIAA0017 |
52.9kb | Anti-sense | Chr. 8 - 110810491 - 110846803 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons |