Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AF251705 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF251705.1 | - | 1 - 2 | 392 | 72 | |
AI060397.1 | - | 1 - 2 | 60 | 72 | ||
AI645367.1 | - | 1 - 2 | 392 | 72 | ||
AK079395.1 | - | 1 - 2 | 120 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |