Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Endov | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164636.1 | NP_001158108.1 | 5 - 6 | 580 | 75 | |
NM_177394.3 | NP_796368.1 | 4 - 5 | 632 | 75 | ||
GenBank mRNA | AK042626.1 | - | 4 - 5 | 520 | 75 | |
AK084233.1 | - | 5 - 6 | 76 | |||
AK084548.1 | - | 5 - 6 | 456 | 75 | ||
AK170595.1 | - | 4 - 5 | 625 | 75 | ||
AK172181.1 | - | 4 - 5 | 599 | 75 | ||
BC056486.1 | - | 4 - 5 | 552 | 5697870 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |