Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogfod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177767.4 | NP_808435.3 | 7 - 8 | 880 | 67 | |
GenBank mRNA | AK029629.1 | - | 7 - 8 | 882 | 67 | |
AK122532.1 | - | 7 - 8 | 784 | 67 | ||
AK156765.1 | - | 7 - 8 | 850 | 67 | ||
AK166343.1 | - | 7 - 8 | 803 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |