Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800008374.mm.1 |
NM_010308.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008374.mm.1 |
NM_010308.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008374.mm.1 |
TC0800001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800008365.mm.1 |
NM_010308.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008365.mm.1 |
NM_010308.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008365.mm.1 |
TC0800001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800008370.mm.1 |
NM_010308.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008370.mm.1 |
NM_010308.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008370.mm.1 |
TC0800001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800008368.mm.1 |
NM_010308.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008368.mm.1 |
NM_010308.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008368.mm.1 |
TC0800001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800008371.mm.1 |
NM_010308.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008371.mm.1 |
NM_010308.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008371.mm.1 |
TC0800001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800008372.mm.1 |
NM_010308.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008372.mm.1 |
NM_010308.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008372.mm.1 |
TC0800001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800008366.mm.1 |
NM_010308.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008366.mm.1 |
NM_010308.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008366.mm.1 |
TC0800001085.mm.1 |
MTA TC |