Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175419.4 | NP_780628.3 | 4 - 5 | 1226 | 56 | |
GenBank mRNA | AK046312.1 | - | 4 - 5 | 1252 | 56 | |
AK142980.1 | - | 4 - 5 | 1228 | 56 | ||
BC052039.1 | - | 4 - 5 | 1207 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |