Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200019780.mm.1 |
NM_145635.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019780.mm.1 |
NM_145635.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019780.mm.1 |
TC0200002435.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019783.mm.1 |
NM_145635.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019783.mm.1 |
NM_145635.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019783.mm.1 |
TC0200002435.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019784.mm.1 |
NM_145635.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019784.mm.1 |
NM_145635.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019784.mm.1 |
TC0200002435.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019787.mm.1 |
NM_145635.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019787.mm.1 |
NM_145635.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019787.mm.1 |
TC0200002435.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019782.mm.1 |
NM_145635.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019782.mm.1 |
NM_145635.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019782.mm.1 |
TC0200002435.mm.1 |
MTA TC |