Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242558.1 | NP_001229487.1 | 5 - 6 | 789 | 68 | |
NM_019825.3 | NP_062799.2 | 5 - 6 | 789 | 68 | ||
GenBank mRNA | AF216186.1 | - | 4 - 5 | 680 | 68 | |
AK144338.1 | - | 7 - 8 | 1028 | 68 | ||
AK171864.1 | - | 5 - 6 | 950 | 68 | ||
BC031113.1 | - | 5 - 6 | 793 | 68 | ||
BC083187.1 | - | 5 - 6 | 789 | 30544739 | 68 | |
JF707634.1 | - | 4 - 5 | 648 | 68 | ||
JF707635.1 | - | 4 - 5 | 648 | 68 | ||
JF707636.1 | - | 4 - 5 | 648 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |