Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085378.2 | NP_001078847.1 | 21 - 22 | 2275 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |