Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026012.2 | NP_080288.1 | 3 - 4 | 420 | 64 | |
GenBank mRNA | AF534394.1 | - | 2 - 3 | 232 | 64 | |
AK007483.1 | - | 1 - 2 | 77 | 64 | ||
AK160222.1 | - | 3 - 4 | 406 | 64 | ||
BC039808.1 | - | 3 - 4 | 390 | 1547029 | 64 | |
BC069856.1 | - | 3 - 4 | 420 | 6491606 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |