Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001144.3 | NP_001001144.2 | 11 - 12 | 1596 | 87 | |
NM_001103162.2 | NP_001096632.1 | 11 - 12 | 1508 | 87 | ||
GenBank mRNA | AK035432.1 | - | 11 - 12 | 1487 | 87 | |
AK146658.1 | - | 11 - 12 | 1578 | 87 | ||
AK150228.1 | - | 10 - 11 | 1512 | 87 | ||
AK150275.1 | - | 11 - 12 | 1480 | 87 | ||
AK152478.1 | - | 11 - 12 | 1480 | 87 | ||
AK153330.1 | - | 11 - 12 | 1480 | 87 | ||
AK172912.1 | - | 10 - 11 | 1988 | 87 | ||
AK202621.1 | - | 3 - 4 | 272 | 87 | ||
BC070437.1 | - | 11 - 12 | 1596 | 30535805 | 87 | |
BC072633.1 | - | 11 - 12 | 1469 | 6849461 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |