Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008694.2 | NP_032720.2 | 1 - 2 | 234 | 83 | |
GenBank mRNA | AK009649.1 | - | 1 - 2 | 188 | 83 | |
AK165210.1 | - | 1 - 2 | 187 | 83 | ||
BC119403.1 | - | 1 - 2 | 205 | 8734152 | 83 | |
BC119405.1 | - | 1 - 2 | 205 | 8734154 | 83 | |
U95002.1 | - | 1 - 2 | 193 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |