Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pth1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083935.1 | NP_001077404.1 | 2 - 3 | 357 | 78 | |
NM_001083936.1 | NP_001077405.1 | 1 - 2 | 390 | 78 | ||
NM_011199.2 | NP_035329.2 | 3 - 4 | 359 | 78 | ||
GenBank mRNA | BB614354.1 | - | 1 - 2 | 391 | 78 | |
BC013446.1 | - | 3 - 4 | 336 | 4241234 | 78 | |
BC051981.1 | - | 1 - 2 | 261 | 6402767 | 78 | |
CB192926.1 | - | 2 - 3 | 319 | 78 | ||
X78936.1 | - | 1 - 2 | 136 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |