Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170684.2 | NP_733785.1 | 10 - 11 | 1179 | 84 | |
GenBank mRNA | AK032541.1 | - | 1 - 2 | 87 | 84 | |
AK049823.1 | - | 7 - 8 | 2321 | 84 | ||
AK162900.1 | - | 9 - 10 | 1798 | 84 | ||
BC119328.1 | - | 10 - 11 | 1078 | 8734077 | 84 | |
BC137595.1 | - | 10 - 11 | 1078 | 8860614 | 84 | |
BM946575.1 | - | 3 - 4 | 242 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |