Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144932.3 | NP_659181.2 | 4 - 5 | 1089 | 60 | |
GenBank mRNA | AK144352.1 | - | 5 - 6 | 1221 | 60 | |
BC022709.1 | - | 3 - 4 | 429 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |