Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301204.1 | NP_001288133.1 | 1 - 2 | 126 | 69 | |
GenBank mRNA | AK089244.1 | - | 1 - 2 | 112 | 69 | |
AK155099.1 | - | 1 - 2 | 78 | 69 | ||
AK155308.1 | - | 1 - 2 | 78 | 69 | ||
AK155417.1 | - | 1 - 2 | 81 | 69 | ||
AK170094.1 | - | 1 - 2 | 78 | 69 | ||
AK170970.1 | - | 1 - 2 | 78 | 69 | ||
AK171044.1 | - | 1 - 2 | 78 | 69 | ||
AK171122.1 | - | 1 - 2 | 78 | 69 | ||
AK171196.1 | - | 1 - 2 | 78 | 69 | ||
AK171268.1 | - | 1 - 2 | 78 | 69 | ||
AK171503.1 | - | 1 - 2 | 78 | 69 | ||
BY209853.1 | - | 1 - 2 | 126 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Mus musculus Probe spans exons |