Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010575.2 | NP_034705.2 | 7 - 8 | 961 | 89 | |
GenBank mRNA | AF166384.1 | - | 7 - 8 | 839 | 89 | |
AF170316.1 | - | 7 - 8 | 822 | 89 | ||
AK048700.1 | - | 3 - 4 | 316 | 89 | ||
AK142289.1 | - | 7 - 8 | 823 | 89 | ||
AK154619.1 | - | 7 - 8 | 962 | 89 | ||
AK158443.1 | - | 7 - 8 | 808 | 89 | ||
BC120493.1 | - | 7 - 8 | 811 | 40129416 | 89 | |
BC137570.1 | - | 7 - 8 | 811 | 8860589 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |