Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145137.2 | NP_660119.1 | 1 - 2 | 87 | 107 | |
GenBank mRNA | BY198732.1 | - | 1 - 2 | 89 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |