Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172371.3 | NP_758959.1 | 1 - 2 | 517 | 97 | |
GenBank mRNA | AK028759.1 | - | 1 - 2 | 518 | 97 | |
BC039949.1 | - | 1 - 2 | 305 | 4167719 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |