Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007528.3 | NP_031554.1 | 6 - 7 | 1138 | 79 | |
GenBank mRNA | AB011665.1 | - | 5 - 6 | 1061 | 79 | |
AK032647.1 | - | 1 - 2 | 129 | 79 | ||
AK156733.1 | - | 6 - 7 | 1100 | 79 | ||
BC090998.1 | - | 6 - 7 | 1098 | 6750884 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |