Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1li2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013380.2 | NP_001013398.2 | 5 - 6 | 951 | 93 | |
GenBank mRNA | AK050575.1 | - | 5 - 6 | 921 | 93 | |
AK140743.1 | - | 5 - 6 | 719 | 93 | ||
AK147899.1 | - | 5 - 6 | 718 | 93 | ||
AK152157.1 | - | 5 - 6 | 716 | 93 | ||
AK161434.1 | - | 5 - 6 | 813 | 93 | ||
AK161494.1 | - | 5 - 6 | 813 | 93 | ||
BC058645.1 | - | 5 - 6 | 909 | 6405596 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |