Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161457.1 | NP_001154929.1 | 3 - 4 | 585 | 90 | |
NM_001161458.1 | NP_001154930.1 | 3 - 4 | 585 | 90 | ||
NM_022309.4 | NP_071704.3 | 3 - 4 | 585 | 90 | ||
GenBank mRNA | AK089094.1 | - | 3 - 4 | 586 | 90 | |
AK134449.1 | - | 2 - 3 | 153 | 90 | ||
BC006763.1 | - | 3 - 4 | 353 | 3481800 | 90 | |
BC026749.1 | - | 3 - 4 | 395 | 4016060 | 90 | |
BC040752.1 | - | 3 - 4 | 384 | 4016060 | 90 | |
D14570.1 | - | 3 - 4 | 515 | 90 | ||
D14571.1 | - | 3 - 4 | 515 | 90 | ||
D14572.1 | - | 3 - 4 | 515 | 90 | ||
L03279.1 | - | 3 - 4 | 364 | 90 | ||
L03306.1 | - | 283 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |