Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam65a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081241.2 | NP_001074710.2 | 1 - 2 | 124 | 74 | |
GenBank mRNA | AK052316.1 | - | 1 - 2 | 73 | 74 | |
AK122573.1 | - | 1 - 2 | 98 | 74 | ||
BB852730.1 | - | 1 - 2 | 124 | 74 | ||
BC079880.1 | - | 1 - 2 | 38 | 6852251 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |