Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctcf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181322.3 | NP_851839.1 | 6 - 7 | 1509 | 88 | |
GenBank mRNA | AB221631.1 | - | 4 - 5 | 1200 | 88 | |
AK132514.1 | - | 4 - 5 | 790 | 88 | ||
AK134247.1 | - | 4 - 5 | 790 | 88 | ||
AK140164.1 | - | 6 - 7 | 1497 | 88 | ||
BC046398.1 | - | 5 - 6 | 1233 | 3982679 | 88 | |
BC049131.1 | - | 6 - 7 | 1468 | 6825952 | 88 | |
BC058240.1 | - | 5 - 6 | 1262 | 6307925 | 88 | |
U51037.1 | - | 6 - 7 | 1506 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |