Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025894.2 | NP_080170.1 | 3 - 4 | 348 | 82 | |
GenBank mRNA | AK005114.1 | - | 3 - 4 | 383 | 82 | |
AK007619.1 | - | 3 - 4 | 393 | 82 | ||
AK007680.1 | - | 3 - 4 | 322 | 82 | ||
AK008646.1 | - | 3 - 4 | 349 | 82 | ||
AK009969.1 | - | 3 - 4 | 351 | 82 | ||
AK011806.1 | - | 3 - 4 | 372 | 82 | ||
AK017843.1 | - | 3 - 4 | 365 | 82 | ||
AK150910.1 | - | 3 - 4 | 372 | 82 | ||
AK151242.1 | - | 3 - 4 | 349 | 82 | ||
AK152041.1 | - | 3 - 4 | 372 | 82 | ||
AK152240.1 | - | 3 - 4 | 380 | 82 | ||
AK160283.1 | - | 3 - 4 | 370 | 82 | ||
AK162779.1 | - | 349 | 82 | |||
AK167236.1 | - | 3 - 4 | 356 | 82 | ||
BC004694.1 | - | 3 - 4 | 366 | 3498952 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |