Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026844.3 | NP_081120.2 | 2 - 3 | 192 | 74 | |
GenBank mRNA | AK002429.1 | - | 2 - 3 | 192 | 74 | |
AK013475.1 | - | 2 - 3 | 189 | 74 | ||
BC027549.1 | - | 2 - 3 | 162 | 1548292 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |