Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atmin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177700.4 | NP_808368.3 | 1 - 2 | 362 | 82 | |
GenBank mRNA | AK122281.1 | - | 1 - 2 | 244 | 82 | |
AX775769.1 | - | 1 - 2 | 362 | 82 | ||
BC027752.1 | - | 1 - 2 | 336 | 82 | ||
BC060631.1 | - | 1 - 2 | 347 | 6847850 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons |