Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200018594.mm.1 |
NM_001285446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018594.mm.1 |
NM_009823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018594.mm.1 |
NM_009823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018594.mm.1 |
NM_172860.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018594.mm.1 |
NM_172860.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018594.mm.1 |
TC0200002341.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018595.mm.1 |
NM_001285446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018595.mm.1 |
NM_009823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018595.mm.1 |
NM_009823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018595.mm.1 |
NM_172860.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018595.mm.1 |
NM_172860.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018595.mm.1 |
TC0200002341.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018593.mm.1 |
NM_001285446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018593.mm.1 |
NM_009823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018593.mm.1 |
NM_009823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018593.mm.1 |
NM_172860.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018593.mm.1 |
NM_172860.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018593.mm.1 |
TC0200002341.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018597.mm.1 |
NM_001285446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018597.mm.1 |
NM_009823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018597.mm.1 |
NM_009823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018597.mm.1 |
NM_172860.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018597.mm.1 |
NM_172860.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018597.mm.1 |
TC0200002341.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018598.mm.1 |
NM_001285446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018598.mm.1 |
NM_009823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018598.mm.1 |
NM_009823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018598.mm.1 |
NM_172860.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018598.mm.1 |
NM_172860.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018598.mm.1 |
TC0200002341.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018584.mm.1 |
NM_001285446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018584.mm.1 |
NM_009823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018584.mm.1 |
NM_009823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018584.mm.1 |
NM_172860.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018584.mm.1 |
NM_172860.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018584.mm.1 |
TC0200002341.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018590.mm.1 |
NM_001285446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018590.mm.1 |
NM_009823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018590.mm.1 |
NM_009823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018590.mm.1 |
NM_172860.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018590.mm.1 |
NM_172860.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018590.mm.1 |
TC0200002341.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018588.mm.1 |
NM_001285446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018588.mm.1 |
NM_009823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018588.mm.1 |
NM_009823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018588.mm.1 |
NM_172860.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018588.mm.1 |
NM_172860.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018588.mm.1 |
TC0200002341.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018592.mm.1 |
NM_001285446.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018592.mm.1 |
NM_009823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018592.mm.1 |
NM_009823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018592.mm.1 |
NM_172860.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018592.mm.1 |
NM_172860.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018592.mm.1 |
TC0200002341.mm.1 |
MTA TC |