Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbfa2t2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285446.1 | NP_001272375.1 | 9 - 10 | 1410 | 67 | |
NM_172860.2 | NP_766448.1 | 9 - 10 | 1414 | 67 | ||
GenBank mRNA | AK049206.1 | - | 9 - 10 | 1372 | 67 | |
AK153602.1 | - | 9 - 10 | 1316 | 67 | ||
BC064679.1 | - | 9 - 10 | 1380 | 6405896 | 67 | |
BC138410.1 | - | 9 - 10 | 1347 | 8861430 | 67 | |
BC145266.1 | - | 9 - 10 | 1343 | 67 | ||
BC145679.1 | - | 9 - 10 | 1347 | 40130982 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |