Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081151.1 | NP_001074620.1 | 4 - 5 | 848 | 68 | |
GenBank mRNA | AK039455.1 | - | 4 - 5 | 847 | 68 | |
BC112432.1 | - | 4 - 5 | 847 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |