Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172913.3 | NP_766501.2 | 1 - 2 | 384 | 83 | |
GenBank mRNA | BC052044.1 | - | 1 - 2 | 110 | 5708447 | 83 |
CK619337.1 | - | 1 - 2 | 475 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |