Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172674.2 | NP_766262.2 | 3 - 4 | 437 | 79 | |
GenBank mRNA | AK033017.1 | - | 2 - 3 | 236 | 79 | |
AK038573.1 | - | 3 - 4 | 344 | 79 | ||
AK045309.1 | - | 3 - 4 | 356 | 79 | ||
BC060121.1 | - | 3 - 4 | 317 | 6413021 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |