Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277186.1 | NP_001264115.1 | 3 - 4 | 1419 | 95 | |
NM_001277187.1 | NP_001264116.1 | 3 - 4 | 1419 | 95 | ||
NM_146128.6 | NP_666240.4 | 3 - 4 | 1419 | 95 | ||
GenBank mRNA | AK122408.1 | - | 3 - 4 | 1419 | 95 | |
AY243849.2 | - | 1 - 2 | 999 | 95 | ||
BC141110.1 | - | 1 - 2 | 1056 | 9055722 | 95 | |
BC145394.1 | - | 1 - 2 | 1056 | 9053952 | 95 | |
BC145395.1 | - | 1 - 2 | 1056 | 9053953 | 95 | |
CX210973.1 | - | 1 - 2 | 203 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |