Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ace3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101453.2 | NP_001094923.2 | 10 - 11 | 1448 | 89 | |
GenBank mRNA | AB531024.1 | - | 10 - 11 | 1448 | 89 | |
CF105940.1 | - | 2 - 3 | 178 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Protease Mus musculus Probe spans exons |