Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ace | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281819.1 | NP_001268748.1 | 14 - 15 | 2272 | 136 | |
NM_009598.2 | NP_033728.1 | 3 - 4 | 495 | 136 | ||
NM_207624.5 | NP_997507.1 | 14 - 15 | 2272 | 136 | ||
GenBank mRNA | AK154632.1 | - | 14 - 15 | 2271 | 136 | |
AK161020.1 | - | 14 - 15 | 2268 | 136 | ||
AK164780.1 | - | 1 - 2 | 140 | 136 | ||
BC034367.1 | - | 10 - 11 | 1534 | 136 | ||
BC040404.1 | - | 14 - 15 | 2261 | 3965649 | 136 | |
BC083109.1 | - | 14 - 15 | 2050 | 136 | ||
BC110362.1 | - | 14 - 15 | 2258 | 5343978 | 136 | |
J04946.1 | - | 8 - 9 | 1239 | 136 | ||
J04947.1 | - | 1239 | 136 | |||
M55333.1 | - | 3 - 4 | 495 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |