Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027946.3 | NP_082222.1 | 3 - 4 | 665 | 64 | |
GenBank mRNA | AK005902.1 | - | 3 - 4 | 659 | 64 | |
AK047435.1 | - | 3 - 4 | 667 | 64 | ||
AK080823.1 | - | 3 - 4 | 639 | 64 | ||
AK154757.1 | - | 3 - 4 | 656 | 64 | ||
AK165674.1 | - | 3 - 4 | 656 | 64 | ||
BC048165.1 | - | 3 - 4 | 581 | 6409531 | 64 | |
BC048722.1 | - | 3 - 4 | 584 | 6504571 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |