Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044744.1 | NP_001038209.2 | 4 - 5 | 476 | 110 | |
NM_008097.2 | NP_032123.3 | 3 - 4 | 533 | 110 | ||
GenBank mRNA | AK078866.1 | - | 1 - 2 | 86 | 110 | |
AK165406.1 | - | 3 - 4 | 359 | 110 | ||
AU080952.1 | - | 4 - 5 | 333 | 110 | ||
BC061158.1 | - | 3 - 4 | 363 | 30314273 | 110 | |
BG807173.1 | - | 3 - 4 | 529 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |