Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010062.3 | NP_034192.1 | 3 - 4 | 505 | 86 | |
GenBank mRNA | AK145192.1 | - | 3 - 4 | 506 | 86 | |
AK167277.1 | - | 3 - 4 | 471 | 86 | ||
BC058609.1 | - | 3 - 4 | 452 | 5397016 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |