Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007398.4 | NP_031424.1 | 11 - 12 | 1225 | 88 | |
GenBank mRNA | AK075899.1 | - | 11 - 12 | 1164 | 88 | |
AK167489.1 | - | 11 - 12 | 1164 | 88 | ||
M10319.1 | - | 11 - 12 | 1147 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Probe spans exons |