Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114079.2 | NP_001107551.1 | 7 - 8 | 1228 | 65 | |
GenBank mRNA | BC158030.1 | - | 7 - 8 | 1228 | 9007333 | 65 |
EU669908.1 | - | 7 - 8 | 971 | 65 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | DV656812.1 | - | 342 | Chr. - - [-] on Build | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |