Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045531.1 | NP_001038996.1 | 2 - 3 | 142 | 117 | |
GenBank mRNA | BC147586.1 | - | 2 - 3 | 124 | 9056576 | 117 |
BC147607.1 | - | 2 - 3 | 124 | 9056597 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |