Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008771.3 | NP_032797.3 | 2 - 3 | 456 | 106 | |
GenBank mRNA | AK035304.1 | - | 2 - 3 | 457 | 106 | |
BC015084.1 | - | 2 - 3 | 470 | 4189541 | 106 | |
X84896.1 | - | 2 - 3 | 450 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |