Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029557.1 | NP_083833.1 | 7 - 8 | 641 | 61 | |
GenBank mRNA | AK002758.1 | - | 7 - 8 | 626 | 61 | |
AK089005.1 | - | 7 - 8 | 608 | 61 | ||
BC126953.1 | - | 6 - 7 | 765 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |