Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148953.2 | NP_683755.1 | 4 - 5 | 94 | ||
GenBank mRNA | AK038411.1 | - | 94 | |||
BC127245.1 | - | 4 - 5 | 94 | |||
BC127246.1 | - | 4 - 5 | 1174 | 94 | ||
BC152348.1 | - | 4 - 5 | 1175 | 94 | ||
BC152349.1 | - | 4 - 5 | 1175 | 40104909 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |