Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030206.4 | NP_084482.1 | 2 - 3 | 776 | 68 | |
GenBank mRNA | AJ315163.1 | - | 2 - 3 | 382 | 68 | |
AK019410.1 | - | 2 - 3 | 777 | 68 | ||
AK087408.1 | - | 2 - 3 | 762 | 68 | ||
AK163073.1 | - | 2 - 3 | 527 | 68 | ||
AK172475.1 | - | 2 - 3 | 523 | 68 | ||
BC055040.1 | - | 4 - 5 | 685 | 6491041 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |