Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp90b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011631.1 | NP_035761.1 | 13 - 14 | 1977 | 96 | |
GenBank mRNA | AK031918.1 | - | 9 - 10 | 1329 | 96 | |
AK146885.1 | - | 13 - 14 | 2043 | 96 | ||
AK150811.1 | - | 13 - 14 | 1997 | 96 | ||
AK151413.1 | - | 13 - 14 | 1994 | 96 | ||
AK153550.1 | - | 13 - 14 | 1997 | 96 | ||
AK159974.1 | - | 13 - 14 | 2002 | 96 | ||
AK160093.1 | - | 13 - 14 | 1996 | 96 | ||
AK160827.1 | - | 1417 | 96 | |||
AK165975.1 | - | 13 - 14 | 1997 | 96 | ||
AK168952.1 | - | 13 - 14 | 1997 | 96 | ||
AK169090.1 | - | 13 - 14 | 1997 | 96 | ||
AK184687.1 | - | 1 - 2 | 245 | 96 | ||
AK189410.1 | - | 1 - 2 | 243 | 96 | ||
AK195996.1 | - | 1 - 2 | 222 | 96 | ||
AK197167.1 | - | 1 - 2 | 243 | 96 | ||
AK216485.1 | - | 1 - 2 | 243 | 96 | ||
AK218224.1 | - | 1 - 2 | 243 | 96 | ||
BC010445.1 | - | 13 - 14 | 1909 | 3587178 | 96 | |
BC011439.1 | - | 13 - 14 | 1983 | 3707817 | 96 | |
J03297.1 | - | 13 - 14 | 1977 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |