Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010578.2 | NP_034708.1 | 2 - 3 | 370 | 97 | |
GenBank mRNA | AK088016.1 | - | 2 - 3 | 271 | 97 | |
AK088729.1 | - | 2 - 3 | 117 | 97 | ||
AK135521.1 | - | 2 - 3 | 269 | 97 | ||
AK159689.1 | - | 2 - 3 | 212 | 97 | ||
AK167682.1 | - | 2 - 3 | 268 | 97 | ||
BC050906.1 | - | 2 - 3 | 243 | 5721348 | 97 | |
X15202.1 | - | 1 - 2 | 63 | 97 | ||
Y00769.1 | - | 2 - 3 | 158 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |